missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl Phosphate Di(tris) Salt Hydrate 90.0+%, TCI America™

Catalog No. N04225G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N04225G Supplier TCI America Supplier No. N04225G
Only null left

Chemical Identifiers

CAS 68189-42-4
Molecular Formula C14H28N3O12P
Molecular Weight (g/mol) 461.361
MDL Number MFCD00037009
InChI Key XXAXKCWOTRABOW-UHFFFAOYSA-N
Synonym Phosphoric Acid 4-Nitrophenyl Ester Bis[tris(hydroxymethyl)aminomethane] Salt, 4-Nitrophenyl Phosphate Bis(tris) Salt
PubChem CID 109721
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;(4-nitrophenyl) dihydrogen phosphate
SMILES C1=CC(=CC=C1[N+](=O)[O-])OP(=O)(O)O.C(C(CO)(CO)N)O.C(C(CO)(CO)N)O

Specifications

Quantity 5 g
Formula Weight 461.36
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenyl Phosphate Di(tris) Salt Hydrate [Substrate for Phosphatase]
TSCA Yes
Recommended Storage Freezer