missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl octanoate, 96%

Catalog No. AAL1202203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1202203 1 g
AAL1202206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1202203 Supplier Thermo Scientific Chemicals Supplier No. L1202203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1956-10-1 | C14H19NO4 | 265.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1956-10-1
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD00024665
InChI Key GGIDEJQGAZSTES-UHFFFAOYSA-N
ChEBI CHEBI:50182

Specifications

Density 1.082
Refractive Index 1.512
Quantity 1 g
Synonym 3,3'-diindolylmethane, di 1h-indol-3-yl methane, diindolylmethane, 3,3'-methylenediindole, 3,3'-methylenebis-1h-indole, 1h-indole, 3,3'-methylenebis, dim, 3-diindolyl methane, arundine, 3,3'-methylenebis 1h-indole
SMILES CCCCCCCC(=O)OC1=CC=C(C=C1)[N+]([O-])=O
IUPAC Name 4-nitrophenyl octanoate
PubChem CID 3071
Formula Weight 265.31
Percent Purity 97%
Chemical Name or Material 4-Nitrophenyl octanoate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only