missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl Isocyanate (contains varying amounts of polymers) 98.0+%, TCI America™

Catalog No. p-7131223
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I01295G 5 g
I012925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I01295G Supplier TCI America Supplier No. I01295G
Only null left

Chemical Identifiers

CAS 100-28-7
Molecular Formula C7H4N2O3
Molecular Weight (g/mol) 164.12
MDL Number MFCD00007306
InChI Key GFNKTLQTQSALEJ-UHFFFAOYSA-N
Synonym 4-nitrophenyl isocyanate, p-nitrophenyl isocyanate, benzene, 1-isocyanato-4-nitro, 4-nitrophenylisocyanate, p-nitrophenylisocyanate, isocyanic acid, p-nitrophenyl ester, 1-isocyanato-4-nitro-benzene, isocyanic acid 4-nitrophenyl ester, timtec-bb sbb006628, 4-nitrobenzenisocyanate
PubChem CID 66012
IUPAC Name 1-isocyanato-4-nitrobenzene
SMILES C1=CC(=CC=C1N=C=O)[N+](=O)[O-]

Specifications

Melting Point 58°C
Color Yellow
Boiling Point 162°C
Quantity 5 g
Formula Weight 164.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenyl Isocyanate (contains varying amounts of polymers)
RTECSNumber DA3692500
TSCA Yes
Recommended Storage Refrigerator