missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl isocyanate, 97%

Catalog No. AAA1119406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1119406 5 g
AAA1119414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1119406 Supplier Thermo Scientific Chemicals Supplier No. A1119406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 100-28-7 | C7H4N2O3 | 164.12 g/mol

4-Nitrophenyl isocyanate is used as pharmaceutical intermediates. 4-Nitrophenyl isocyanate was used in the synthesis of 2-(5-methyl-3,4-diphenyl-1H-pyrrole-2-carbonyl)-N-(4-nitrophenyl)hydrazinecarboxamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl isocyanate is used as pharmaceutical intermediates. 4-Nitrophenyl isocyanate was used in the synthesis of 2-(5-methyl-3,4-diphenyl-1H-pyrrole-2-carbonyl)-N-(4-nitrophenyl)hydrazinecarboxamide.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store at 2-8°C. Incompatible with water, heat and oxidizing agents.

Chemical Identifiers

CAS 100-28-7
Molecular Formula C7H4N2O3
Molecular Weight (g/mol) 164.12
MDL Number MFCD00007306
InChI Key GFNKTLQTQSALEJ-UHFFFAOYSA-N
Synonym 4-nitrophenyl isocyanate, p-nitrophenyl isocyanate, benzene, 1-isocyanato-4-nitro, 4-nitrophenylisocyanate, p-nitrophenylisocyanate, isocyanic acid, p-nitrophenyl ester, 1-isocyanato-4-nitro-benzene, isocyanic acid 4-nitrophenyl ester, timtec-bb sbb006628, 4-nitrobenzenisocyanate
PubChem CID 66012
IUPAC Name 1-isocyanato-4-nitrobenzene
SMILES C1=CC(=CC=C1N=C=O)[N+](=O)[O-]

Specifications

Melting Point 56°C to 59°C
Boiling Point 137°C to 138°C (11 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3335
Beilstein 389516
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 164.12
Percent Purity 97%
Chemical Name or Material 4-Nitrophenyl isocyanate
Show More Show Less
Hazard Category H302+H332-H315-H319-H334-H335
Hazard Statement GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P332+P313-P342+P311-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 202-836-3
RTECSNumber DA3692500
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only