missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl-beta-D-glucopyranoside, 99%

Catalog No. AC303110250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC303110250 25 g Glass bottle
AC303110010 1 g Glass bottle
AC303110050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC303110250 Supplier Thermo Scientific Chemicals Supplier No. 303110250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2492-87-7 | C12H15NO8 | 301.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2492-87-7
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.25
MDL Number MFCD00006593
InChI Key IFBHRQDFSNCLOZ-RMPHRYRLSA-N
Synonym 4-nitrophenyl-beta-d-glucopyranoside, pnpg, 4-nitrophenyl beta-d-glucopyranoside, 1-o-p-nitrophenyl-d-glucose, 4-nitrophenyl beta-d-glucoside, p-nitrophenyl beta-d-glucopyranoside, p-nitrophenyl beta-d-glucoside, glucoside, p-nitrophenyl, p-nitrophenyl-beta-glucoside, p-nitrophenyl beta-glucoside
PubChem CID 92930
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 165.0°C to 168.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 25 g
Specific Rotation Condition − 107.50 (20.00°C c=1,H2O)
Specific Rotation − 107.50
Formula Weight 301.25
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Nitrophenyl-β-D-glucopyranoside
Show More Show Less
EINECSNumber 219-661-3

RUO – Research Use Only