missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenyl beta-D-Glucuronide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 93°C |
| Color | White-Yellow |
| Quantity | 100 mg |
| Formula Weight | 315.23 |
| Percent Purity | ≥98.0% (HPLC) |
| Grade | Research |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Nitrophenyl beta-D-Glucuronide [Substrate for beta-Glucuronidase] |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |