missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl beta-D-Glucopyranoside Monohydrate 98.0+%, TCI America™

Catalog No. N02351G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02351G 1 g
N02355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N02351G Supplier TCI America Supplier No. N02351G

Chemical Identifiers

CAS 2492-87-7
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.25
MDL Number MFCD00006593
InChI Key IFBHRQDFSNCLOZ-RMPHRYRLSA-N
Synonym 4-nitrophenyl-beta-d-glucopyranoside, pnpg, 4-nitrophenyl beta-d-glucopyranoside, 1-o-p-nitrophenyl-d-glucose, 4-nitrophenyl beta-d-glucoside, p-nitrophenyl beta-d-glucopyranoside, p-nitrophenyl beta-d-glucoside, glucoside, p-nitrophenyl, p-nitrophenyl-beta-glucoside, p-nitrophenyl beta-glucoside
PubChem CID 92930
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 167°C
Color White-Yellow
Quantity 1 g
Formula Weight 301.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenyl beta-D-Glucopyranoside Monohydrate [Substrate for beta-D-Glucosidase]
TSCA Yes
Recommended Storage Refrigerator