missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl-beta-D-glucopyranoside, 98+%

CAS: 2492-87-7 | C12H15NO8 | 301.25 g/mol

$165.74 - $602.37

Chemical Identifiers

CAS 2492-87-7
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.25
MDL Number MFCD00006593
InChI Key IFBHRQDFSNCLOZ-RMPHRYRLSA-N
Synonym 4-nitrophenyl-beta-d-glucopyranoside, pnpg, 4-nitrophenyl beta-d-glucopyranoside, 1-o-p-nitrophenyl-d-glucose, 4-nitrophenyl beta-d-glucoside, p-nitrophenyl beta-d-glucopyranoside, p-nitrophenyl beta-d-glucoside, glucoside, p-nitrophenyl, p-nitrophenyl-beta-glucoside, p-nitrophenyl beta-glucoside
PubChem CID 92930
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAL1015403
View Documents Promotion Details
N/A 1 g
Each for $165.74
Only null left
 
AAL1015406
View Documents Promotion Details
N/A 5 g
Each for $602.37
Only null left
 
Description

Description

4-Nitrophenyl-β-D-glucopyranoside acts as a chromogenic beta-D-glucosidase substrate used for the measurement of beta-glucosidase activity. Further, it is used in research, biochemical enzyme assays and in vitro diagnostic analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl-β-D-glucopyranoside acts as a chromogenic beta-D-glucosidase substrate used for the measurement of beta-glucosidase activity. Further, it is used in research, biochemical enzyme assays and in vitro diagnostic analysis.

Solubility
Soluble in water, methanol, warm ethanol, dimethyl sulfoxide and dimethylformamide. Insoluble in ether.

Notes
Store in a cool place. Incompatible with strong bases.
Specifications

Chemical Identifiers

2492-87-7
301.25
IFBHRQDFSNCLOZ-RMPHRYRLSA-N
92930
OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O
C12H15NO8
MFCD00006593
4-nitrophenyl-beta-d-glucopyranoside, pnpg, 4-nitrophenyl beta-d-glucopyranoside, 1-o-p-nitrophenyl-d-glucose, 4-nitrophenyl beta-d-glucoside, p-nitrophenyl beta-d-glucopyranoside, p-nitrophenyl beta-d-glucoside, glucoside, p-nitrophenyl, p-nitrophenyl-beta-glucoside, p-nitrophenyl beta-glucoside
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol

Specifications

2492-87-7
C12H15NO8
92211
Soluble in water,methanol,warm ethanol,dimethyl sulfoxide and dimethylformamide. Insoluble in ether.
OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
92930
≥98%
166°C to 167°C
MFCD00006593
4-nitrophenyl-beta-d-glucopyranoside, pnpg, 4-nitrophenyl beta-d-glucopyranoside, 1-o-p-nitrophenyl-d-glucose, 4-nitrophenyl beta-d-glucoside, p-nitrophenyl beta-d-glucopyranoside, p-nitrophenyl beta-d-glucoside, glucoside, p-nitrophenyl, p-nitrophenyl-beta-glucoside, p-nitrophenyl beta-glucoside
IFBHRQDFSNCLOZ-RMPHRYRLSA-N
−103° (c=1 in Water)
301.25
301.25
4-Nitrophenyl-beta-D-glucopyranoside
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 219-661-3

TSCA : Yes

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only