missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenyl beta-D-Galactopyranoside 98.0+%, TCI America™
Chemical Identifiers
| CAS | 3150-24-1 |
|---|---|
| Molecular Formula | C12H15NO8 |
| Molecular Weight (g/mol) | 301.25 |
| MDL Number | MFCD00063256 |
| InChI Key | IFBHRQDFSNCLOZ-YBXAARCKSA-N |
| Synonym | 4-nitrophenyl-beta-d-galactopyranoside, 4-nitrophenylgalactoside, p-nitrophenyl-beta-d-galactoside, p-nitrophenyl beta-d-galactopyranoside, 4-nitrophenyl beta-d-galactopyranoside, p-nitrophenol galactoside, 4-nitrophenyl-beta-d-galactoside, 4-nitrophenyl galactoside, 1-o-p-nitrophenyl-beta-d-galactopyranose, 4-nitrophenylbeta-d-galactopyranoside |
| PubChem CID | 65115 |
| ChEBI | CHEBI:355715 |
| IUPAC Name | (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol |
| SMILES | OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O |
Chemical Identifiers
| 3150-24-1 | |
| 301.25 | |
| IFBHRQDFSNCLOZ-YBXAARCKSA-N | |
| 65115 | |
| (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol |
| C12H15NO8 | |
| MFCD00063256 | |
| 4-nitrophenyl-beta-d-galactopyranoside, 4-nitrophenylgalactoside, p-nitrophenyl-beta-d-galactoside, p-nitrophenyl beta-d-galactopyranoside, 4-nitrophenyl beta-d-galactopyranoside, p-nitrophenol galactoside, 4-nitrophenyl-beta-d-galactoside, 4-nitrophenyl galactoside, 1-o-p-nitrophenyl-beta-d-galactopyranose, 4-nitrophenylbeta-d-galactopyranoside | |
| CHEBI:355715 | |
| OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O |
Specifications
| 3150-24-1 | |
| White-Yellow | |
| MFCD00063256 | |
| IFBHRQDFSNCLOZ-YBXAARCKSA-N | |
| (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol | |
| 65115 | |
| 301.25 | |
| Crystalline Powder |
| 180°C | |
| C12H15NO8 | |
| 4-nitrophenyl-beta-d-galactopyranoside, 4-nitrophenylgalactoside, p-nitrophenyl-beta-d-galactoside, p-nitrophenyl beta-d-galactopyranoside, 4-nitrophenyl beta-d-galactopyranoside, p-nitrophenol galactoside, 4-nitrophenyl-beta-d-galactoside, 4-nitrophenyl galactoside, 1-o-p-nitrophenyl-beta-d-galactopyranose, 4-nitrophenylbeta-d-galactopyranoside | |
| OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O | |
| 301.25 | |
| CHEBI:355715 | |
| ≥98.0% (HPLC) | |
| 4-Nitrophenyl beta-D-Galactopyranoside [Substrate for beta-Galactosidase] |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator