missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl-alpha-D-glucopyranoside, 98+%

Catalog No. AAL1007006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1007006 5 g
AAL1007003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1007006 Supplier Thermo Scientific Chemicals Supplier No. L1007006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3767-28-0 | C12H15NO8 | 301.251 g/mol

4-Nitrophenyl-α-D-glucopyranoside acts as a chromogenic substrate for alfa-D-glucosidase inhibitor. It is also used in the detection of glucansucrases and for yeast alfa-D-glucosidase. Further, it is a substrate for lysosomal alfa-glucosidase and maltase-glucoamylase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl-α-D-glucopyranoside acts as a chromogenic substrate for alfa-D-glucosidase inhibitor. It is also used in the detection of glucansucrases and for yeast alfa-D-glucosidase. Further, it is a substrate for lysosomal alfa-glucosidase and maltase-glucoamylase.

Solubility
Soluble in water, warm ethanol and methanol.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Light sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 3767-28-0
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.251
MDL Number MFCD00064088
InChI Key IFBHRQDFSNCLOZ-ZIQFBCGOSA-N
Synonym 4-nitrophenyl-alpha-d-glucopyranoside, 4-nitrophenyl alpha-d-glucopyranoside, 4-nitrophenyl a-d-glucopyranoside, 4-nitrophenyl alpha-glucoside, pnpalphaglu, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-4-nitrophenoxy oxane-3,4,5-triol, p-nitrophenyl alpha-d-glucopyranoside, 4'-nitrophenyl-alpha-d-glucopyranoside, png, p-nitrophenyl, a-d-glucopyranoside
PubChem CID 92969
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 206°C to 207°C
Quantity 5 g
Beilstein 92212
Solubility Information Soluble in water,warm ethanol and methanol.
Optical Rotation +218° (c=0.5 in Water)
Formula Weight 301.25
Percent Purity ≥98%
Chemical Name or Material 4-Nitrophenyl-alpha-D-glucopyranoside
EINECSNumber 223-189-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only