missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroindole-3-carboxaldehyde, 96%

Catalog No. p-7055101
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6359503 1 g
AAH6359506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6359503 Supplier Thermo Scientific Chemicals Supplier No. H6359503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10553-11-4 | C9H6N2O3 | 190.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10553-11-4
Molecular Formula C9H6N2O3
Molecular Weight (g/mol) 190.16
MDL Number MFCD04973979
InChI Key CGXVTWQTGQAMMX-UHFFFAOYSA-N
Synonym 4-nitroindole-3-carboxaldehyde, acmc-20amck, 3-formyl-4-nitroindole, 4-nitroindole-3-carbaldehyde, 4-nitro-1h-indol-3-carbaldehyde, 1h-indole-3-carboxaldehyde,4-nitro
PubChem CID 11095397
IUPAC Name 4-nitro-1H-indole-3-carbaldehyde
SMILES [O-][N+](=O)C1=C2C(NC=C2C=O)=CC=C1

Specifications

Melting Point 189°C to 193°C
Color Yellow
Quantity 1 g
Formula Weight 190.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Nitroindole-3-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only