missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroimidazole 98.0+%, TCI America™

Catalog No. N0506500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0506500G 500 g
N050625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0506500G Supplier TCI America Supplier No. N0506500G

Chemical Identifiers

CAS 3034-38-6
Molecular Formula C3H3N3O2
Molecular Weight (g/mol) 113.08
MDL Number MFCD00005196
InChI Key VYDWQPKRHOGLPA-UHFFFAOYSA-N
Synonym 4-nitroimidazole, 4-nitro-1h-imidazole, 1h-imidazole, 4-nitro, 5-nitroimidazole, 4 5-nitroimidazole, imidazole, 4-nitro, 1h-imidazole, 5-nitro, unii-y8u32az5o7, 5 4-nitroimidazole, 4-nitro-3h-imidazole
PubChem CID 18208
ChEBI CHEBI:64635
IUPAC Name 5-nitro-1H-imidazole
SMILES [O-][N+](=O)C1=CN=CN1

Specifications

Melting Point 312°C
Color Yellow
Quantity 500 g
Formula Weight 113.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitroimidazole
RTECSNumber NI7892000
TSCA Yes