missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroimidazole, 98%

Catalog No. AC161600250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161600250 25 g Plastic bottle
AC161601000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC161600250 Supplier Thermo Scientific Chemicals Supplier No. 161600250

CAS: 3034-38-6 | C3H3N3O2 | 113.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3034-38-6
Molecular Formula C3H3N3O2
Molecular Weight (g/mol) 113.08
MDL Number MFCD00005196
InChI Key VYDWQPKRHOGLPA-UHFFFAOYSA-N
Synonym 4-nitroimidazole, 4-nitro-1h-imidazole, 1h-imidazole, 4-nitro, 5-nitroimidazole, 4 5-nitroimidazole, imidazole, 4-nitro, 1h-imidazole, 5-nitro, unii-y8u32az5o7, 5 4-nitroimidazole, 4-nitro-3h-imidazole
PubChem CID 18208
ChEBI CHEBI:64635
IUPAC Name 5-nitro-1H-imidazole
SMILES [O-][N+](=O)C1=CN=CN1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 303.0°C
Color White to Yellow
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 25 g
Beilstein 23, 50
Solubility Information Solubility in water: slightly soluble in water. Other solubilities: soluble in dilute acids,soluble in most common organic solvents
Formula Weight 113.08
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Nitroimidazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 221-224-7

RUO – Research Use Only