missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrocinnamic Acid 99.0+%, TCI America™
$115.57 - $229.29
Chemical Identifiers
| CAS | 619-89-6 |
|---|---|
| Molecular Formula | C9H7NO4 |
| Molecular Weight (g/mol) | 193.158 |
| MDL Number | MFCD00007381 |
| InChI Key | XMMRNCHTDONGRJ-ZZXKWVIFSA-N |
| Synonym | 4-nitrocinnamic acid, 3-4-nitrophenyl acrylic acid, p-nitrocinnamic acid, cinnamic acid, p-nitro, 2-propenoic acid, 3-4-nitrophenyl, e-3-4-nitrophenyl acrylic acid, 3-4-nitrophenyl propenoic acid, trans-4-nitrocinnamic acid, ccris 1665, 2e-3-4-nitrophenyl prop-2-enoic acid |
| PubChem CID | 737157 |
| SMILES | C1=CC(=CC=C1C=CC(=O)O)[N+](=O)[O-] |
Chemical Identifiers
| 619-89-6 | |
| 193.158 | |
| XMMRNCHTDONGRJ-ZZXKWVIFSA-N | |
| 737157 |
| C9H7NO4 | |
| MFCD00007381 | |
| 4-nitrocinnamic acid, 3-4-nitrophenyl acrylic acid, p-nitrocinnamic acid, cinnamic acid, p-nitro, 2-propenoic acid, 3-4-nitrophenyl, e-3-4-nitrophenyl acrylic acid, 3-4-nitrophenyl propenoic acid, trans-4-nitrocinnamic acid, ccris 1665, 2e-3-4-nitrophenyl prop-2-enoic acid | |
| C1=CC(=CC=C1C=CC(=O)O)[N+](=O)[O-] |
Specifications
| 619-89-6 | |
| C9H7NO4 | |
| 10 g | |
| XMMRNCHTDONGRJ-ZZXKWVIFSA-N | |
| 193.158 | |
| 193.16 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00007381 | |
| 4-nitrocinnamic acid, 3-4-nitrophenyl acrylic acid, p-nitrocinnamic acid, cinnamic acid, p-nitro, 2-propenoic acid, 3-4-nitrophenyl, e-3-4-nitrophenyl acrylic acid, 3-4-nitrophenyl propenoic acid, trans-4-nitrocinnamic acid, ccris 1665, 2e-3-4-nitrophenyl prop-2-enoic acid | |
| C1=CC(=CC=C1C=CC(=O)O)[N+](=O)[O-] | |
| 737157 | |
| ≥99.0% (T) | |
| 4-Nitrocinnamic Acid |
Safety and Handling
EINECSNumber : (3)-1705
RTECSNumber : GE0353000
TSCA : Yes