missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrocinnamic Acid 99.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 193.16 |
| Percent Purity | ≥99.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Nitrocinnamic Acid |
Safety and Handling
| EINECSNumber | (3)-1705 |
| RTECSNumber | GE0353000 |
| TSCA | Yes |