missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrocinnamic Acid 99.0+%, TCI America™

Catalog No. N035510G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N035510G 10 g
N035525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N035510G Supplier TCI America Supplier No. N035510G
Only null left

Chemical Identifiers

CAS 619-89-6
Molecular Formula C9H7NO4
Molecular Weight (g/mol) 193.158
MDL Number MFCD00007381
InChI Key XMMRNCHTDONGRJ-ZZXKWVIFSA-N
Synonym 4-nitrocinnamic acid, 3-4-nitrophenyl acrylic acid, p-nitrocinnamic acid, cinnamic acid, p-nitro, 2-propenoic acid, 3-4-nitrophenyl, e-3-4-nitrophenyl acrylic acid, 3-4-nitrophenyl propenoic acid, trans-4-nitrocinnamic acid, ccris 1665, 2e-3-4-nitrophenyl prop-2-enoic acid
PubChem CID 737157
SMILES C1=CC(=CC=C1C=CC(=O)O)[N+](=O)[O-]

Specifications

Color White-Yellow
Quantity 10 g
Formula Weight 193.16
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrocinnamic Acid
EINECSNumber (3)-1705
RTECSNumber GE0353000
TSCA Yes