missing translation for 'onlineSavingsMsg'
Learn More

4'-Nitrobiphenyl-4-carboxaldehyde 98.0+%, TCI America™

Catalog No. N09671G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09671G 1 g
N09675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09671G Supplier TCI America Supplier No. N09671G
Only null left

Chemical Identifiers

CAS 98648-23-8
Molecular Formula C13H9NO3
Molecular Weight (g/mol) 227.22
MDL Number MFCD06200931
InChI Key MGHIIGWDUVNPPV-UHFFFAOYSA-N
PubChem CID 2763586
IUPAC Name 4'-nitro-[1,1'-biphenyl]-4-carbaldehyde
SMILES [O-][N+](=O)C1=CC=C(C=C1)C1=CC=C(C=O)C=C1

Specifications

Melting Point 128°C
Color Yellow
Quantity 1 g
Formula Weight 227.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Nitrobiphenyl-4-carboxaldehyde