missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrobenzylamine Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 265°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 188.61 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Nitrobenzylamine Hydrochloride |
Safety and Handling
| RTECSNumber | DP6705000 |
| TSCA | No |