missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrobenzyl (2S,4S)-2-(Dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylate 95.0+%, TCI America™
$294.52 - $1021.51
Chemical Identifiers
| CAS | 96034-64-9 |
|---|---|
| Molecular Formula | C15H19N3O5S |
| Molecular Weight (g/mol) | 353.393 |
| MDL Number | MFCD08063348 |
| InChI Key | VGLBNJWGUYQZHD-STQMWFEESA-N |
| Synonym | (2S,4S)-2-(Dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylic Acid 4-Nitrobenzyl Ester |
| PubChem CID | 10871907 |
| IUPAC Name | (4-nitrophenyl)methyl (2S,4S)-2-(dimethylcarbamoyl)-4-sulfanylpyrrolidine-1-carboxylate |
| SMILES | CN(C)C(=O)C1CC(CN1C(=O)OCC2=CC=C(C=C2)[N+](=O)[O-])S |
Chemical Identifiers
| 96034-64-9 | |
| 353.393 | |
| VGLBNJWGUYQZHD-STQMWFEESA-N | |
| 10871907 | |
| CN(C)C(=O)C1CC(CN1C(=O)OCC2=CC=C(C=C2)[N+](=O)[O-])S |
| C15H19N3O5S | |
| MFCD08063348 | |
| (2S,4S)-2-(Dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylic Acid 4-Nitrobenzyl Ester | |
| (4-nitrophenyl)methyl (2S,4S)-2-(dimethylcarbamoyl)-4-sulfanylpyrrolidine-1-carboxylate |
Specifications
| 96034-64-9 | |
| White-Yellow | |
| MFCD08063348 | |
| (2S,4S)-2-(Dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylic Acid 4-Nitrobenzyl Ester | |
| CN(C)C(=O)C1CC(CN1C(=O)OCC2=CC=C(C=C2)[N+](=O)[O-])S | |
| 353.393 | |
| 353.39 | |
| Crystalline Powder |
| 119°C | |
| C15H19N3O5S | |
| 1 g | |
| VGLBNJWGUYQZHD-STQMWFEESA-N | |
| (4-nitrophenyl)methyl (2S,4S)-2-(dimethylcarbamoyl)-4-sulfanylpyrrolidine-1-carboxylate | |
| 10871907 | |
| ≥95.0% (HPLC,T) | |
| 4-Nitrobenzyl (2S,4S)-2-(Dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylate |
Safety and Handling
TSCA : No