missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzoyl Chloride 98.0+%, TCI America™

Catalog No. p-7133660
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
N017625G 25 g
N0176500G 500 g
2 options

Catalog No. N017625G

Supplier: TCI America N017625G

Only null left

Chemical Identifiers

CAS 122-04-3
Molecular Formula C7H4ClNO3
Molecular Weight (g/mol) 185.56
MDL Number MFCD00007345
InChI Key SKDHHIUENRGTHK-UHFFFAOYSA-N
Synonym p-nitrobenzoyl chloride, benzoyl chloride, 4-nitro, 4-nitrobenzoic acid chloride, p-nitrobenzoic acid chloride, benzoyl chloride, p-nitro, 4-nitro-benzoyl chloride, unii-x3h8pw2gc4, ccris 3136, 4-nitrobenzoylchloride, 4-nitro benzoyl chloride
PubChem CID 8502
IUPAC Name 4-nitrobenzoyl chloride
SMILES [O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O

Specifications

Melting Point 73°C
Color Yellow
Boiling Point 128°C
Quantity 25 g
UN Number 3261
Formula Weight 185.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzoyl Chloride
EINECSNumber (3)-1504
RTECSNumber DM6651000
TSCA Yes