missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Nitrobenzo-18-crown 6-Ether 97.0+%, TCI America™
$317.66 - $1266.77
Chemical Identifiers
| CAS | 53408-96-1 |
|---|---|
| Molecular Formula | C16H23NO8 |
| Molecular Weight (g/mol) | 357.359 |
| MDL Number | MFCD00143207 |
| InChI Key | LQXOKBZWNFJJGI-UHFFFAOYSA-N |
| Synonym | 4-nitrobenzo-18-crown-6, 4'-nitrobenzo-18-crown 6-ether, nitrobenzo-18-crown-6, 18-nitro-2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine, 20-nitro-2,5,8,11,14,17-hexaoxabicyclo 16.4.0 docosa-1 18 ,19,21-triene, 4'-nitrobenzo-18-crown-6, lqxokbzwnfjjgi-uhfffaoysa, 2,3-4-nitrobenzo-1,4,7,10,13,16-hexaoxacycloocta, 2,3-4-nitrobenzo-1,4,7,10,13,16-hexaoxacyclooctadec-2-ene |
| PubChem CID | 602198 |
| IUPAC Name | 20-nitro-2,5,8,11,14,17-hexaoxabicyclo[16.4.0]docosa-1(18),19,21-triene |
| SMILES | C1COCCOCCOC2=C(C=CC(=C2)[N+](=O)[O-])OCCOCCO1 |
Chemical Identifiers
| 53408-96-1 | |
| 357.359 | |
| LQXOKBZWNFJJGI-UHFFFAOYSA-N | |
| 602198 | |
| C1COCCOCCOC2=C(C=CC(=C2)[N+](=O)[O-])OCCOCCO1 |
| C16H23NO8 | |
| MFCD00143207 | |
| 4-nitrobenzo-18-crown-6, 4'-nitrobenzo-18-crown 6-ether, nitrobenzo-18-crown-6, 18-nitro-2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine, 20-nitro-2,5,8,11,14,17-hexaoxabicyclo 16.4.0 docosa-1 18 ,19,21-triene, 4'-nitrobenzo-18-crown-6, lqxokbzwnfjjgi-uhfffaoysa, 2,3-4-nitrobenzo-1,4,7,10,13,16-hexaoxacycloocta, 2,3-4-nitrobenzo-1,4,7,10,13,16-hexaoxacyclooctadec-2-ene | |
| 20-nitro-2,5,8,11,14,17-hexaoxabicyclo[16.4.0]docosa-1(18),19,21-triene |
Specifications
| 53408-96-1 | |
| Yellow | |
| MFCD00143207 | |
| 4-nitrobenzo-18-crown-6, 4'-nitrobenzo-18-crown 6-ether, nitrobenzo-18-crown-6, 18-nitro-2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine, 20-nitro-2,5,8,11,14,17-hexaoxabicyclo 16.4.0 docosa-1 18 ,19,21-triene, 4'-nitrobenzo-18-crown-6, lqxokbzwnfjjgi-uhfffaoysa, 2,3-4-nitrobenzo-1,4,7,10,13,16-hexaoxacycloocta, 2,3-4-nitrobenzo-1,4,7,10,13,16-hexaoxacyclooctadec-2-ene | |
| C1COCCOCCOC2=C(C=CC(=C2)[N+](=O)[O-])OCCOCCO1 | |
| 357.359 | |
| 357.36 | |
| Crystalline Powder |
| 86°C | |
| C16H23NO8 | |
| 1 g | |
| LQXOKBZWNFJJGI-UHFFFAOYSA-N | |
| 20-nitro-2,5,8,11,14,17-hexaoxabicyclo[16.4.0]docosa-1(18),19,21-triene | |
| 602198 | |
| ≥97.0% (GC) | |
| 4′-Nitrobenzo-18-crown 6-Ether |
Safety and Handling
TSCA : No