missing translation for 'onlineSavingsMsg'
Learn More

4'-Nitrobenzo-18-crown 6-Ether 97.0+%, TCI America™

Catalog No. N05625G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05625G 5 g
N05621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05625G Supplier TCI America Supplier No. N05625G

Chemical Identifiers

CAS 53408-96-1
Molecular Formula C16H23NO8
Molecular Weight (g/mol) 357.359
MDL Number MFCD00143207
InChI Key LQXOKBZWNFJJGI-UHFFFAOYSA-N
Synonym 4-nitrobenzo-18-crown-6, 4'-nitrobenzo-18-crown 6-ether, nitrobenzo-18-crown-6, 18-nitro-2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine, 20-nitro-2,5,8,11,14,17-hexaoxabicyclo 16.4.0 docosa-1 18 ,19,21-triene, 4'-nitrobenzo-18-crown-6, lqxokbzwnfjjgi-uhfffaoysa, 2,3-4-nitrobenzo-1,4,7,10,13,16-hexaoxacycloocta, 2,3-4-nitrobenzo-1,4,7,10,13,16-hexaoxacyclooctadec-2-ene
PubChem CID 602198
IUPAC Name 20-nitro-2,5,8,11,14,17-hexaoxabicyclo[16.4.0]docosa-1(18),19,21-triene
SMILES C1COCCOCCOC2=C(C=CC(=C2)[N+](=O)[O-])OCCOCCO1

Specifications

Melting Point 86°C
Color Yellow
Quantity 5 g
Formula Weight 357.36
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Nitrobenzo-18-crown 6-Ether