missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Nitrobenzo-18-crown 6-Ether 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 86°C |
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 357.36 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4′-Nitrobenzo-18-crown 6-Ether |
Safety and Handling
| TSCA | No |