missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzo-18-crown-6, 99%

Catalog No. AAA1613806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1613806 5 g
AAA1613803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1613806 Supplier Thermo Scientific Chemicals Supplier No. A1613806

CAS: 53408-96-1 | C16H23NO8 | 357.359 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53408-96-1
Molecular Formula C16H23NO8
Molecular Weight (g/mol) 357.359
MDL Number MFCD00143207
InChI Key LQXOKBZWNFJJGI-UHFFFAOYSA-N
Synonym 4-nitrobenzo-18-crown-6, 4'-nitrobenzo-18-crown 6-ether, nitrobenzo-18-crown-6, 18-nitro-2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine, 20-nitro-2,5,8,11,14,17-hexaoxabicyclo 16.4.0 docosa-1 18 ,19,21-triene, 4'-nitrobenzo-18-crown-6, lqxokbzwnfjjgi-uhfffaoysa, 2,3-4-nitrobenzo-1,4,7,10,13,16-hexaoxacycloocta, 2,3-4-nitrobenzo-1,4,7,10,13,16-hexaoxacyclooctadec-2-ene
PubChem CID 602198
IUPAC Name 20-nitro-2,5,8,11,14,17-hexaoxabicyclo[16.4.0]docosa-1(18),19,21-triene
SMILES C1COCCOCCOC2=C(C=CC(=C2)[N+](=O)[O-])OCCOCCO1

Specifications

Melting Point 84°C to 86°C
Quantity 5 g
Formula Weight 357.35
Percent Purity 99%
Chemical Name or Material 4-Nitrobenzo-18-crown-6
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only