missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Nitrobenzo-15-crown 5-Ether 99.0+%, TCI America™
$128.62 - $387.73
Chemical Identifiers
| CAS | 60835-69-0 |
|---|---|
| Molecular Formula | C14H19NO7 |
| Molecular Weight (g/mol) | 313.306 |
| MDL Number | MFCD00060713 |
| InChI Key | XIWRBQVYCZCEPG-UHFFFAOYSA-N |
| Synonym | 4-nitrobenzo-15-crown-5, 4'-nitrobenzo-15-crown-5, nitrobenzo-15-crown-5, 15-nitro-2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine, 17-nitro-2,5,8,11,14-pentaoxabicyclo 13.4.0 nonadeca-1 15 ,16,18-triene, 17-nitro-2,3-benzo-1,4,7,10,13-pentaoxacyclopentadecen-2, 4-nitro-benzo-15-crown-5, xiwrbqvyczcepg-uhfffaoysa, 4'-nitrobenzo-15-crown 5-ether |
| PubChem CID | 143747 |
| IUPAC Name | 17-nitro-2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene |
| SMILES | C1COCCOC2=C(C=C(C=C2)[N+](=O)[O-])OCCOCCO1 |
Chemical Identifiers
| 60835-69-0 | |
| 313.306 | |
| XIWRBQVYCZCEPG-UHFFFAOYSA-N | |
| 143747 | |
| C1COCCOC2=C(C=C(C=C2)[N+](=O)[O-])OCCOCCO1 |
| C14H19NO7 | |
| MFCD00060713 | |
| 4-nitrobenzo-15-crown-5, 4'-nitrobenzo-15-crown-5, nitrobenzo-15-crown-5, 15-nitro-2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine, 17-nitro-2,5,8,11,14-pentaoxabicyclo 13.4.0 nonadeca-1 15 ,16,18-triene, 17-nitro-2,3-benzo-1,4,7,10,13-pentaoxacyclopentadecen-2, 4-nitro-benzo-15-crown-5, xiwrbqvyczcepg-uhfffaoysa, 4'-nitrobenzo-15-crown 5-ether | |
| 17-nitro-2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene |
Specifications
| 60835-69-0 | |
| Yellow | |
| MFCD00060713 | |
| 4-nitrobenzo-15-crown-5, 4'-nitrobenzo-15-crown-5, nitrobenzo-15-crown-5, 15-nitro-2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine, 17-nitro-2,5,8,11,14-pentaoxabicyclo 13.4.0 nonadeca-1 15 ,16,18-triene, 17-nitro-2,3-benzo-1,4,7,10,13-pentaoxacyclopentadecen-2, 4-nitro-benzo-15-crown-5, xiwrbqvyczcepg-uhfffaoysa, 4'-nitrobenzo-15-crown 5-ether | |
| C1COCCOC2=C(C=C(C=C2)[N+](=O)[O-])OCCOCCO1 | |
| 313.306 | |
| 313.31 | |
| Crystalline Powder |
| 98°C | |
| C14H19NO7 | |
| 1 g | |
| XIWRBQVYCZCEPG-UHFFFAOYSA-N | |
| 17-nitro-2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene | |
| 143747 | |
| ≥99.0% (GC) | |
| 4′-Nitrobenzo-15-crown 5-Ether |
Safety and Handling
TSCA : No