missing translation for 'onlineSavingsMsg'
Learn More

4'-Nitrobenzo-15-crown 5-Ether 99.0+%, TCI America™

Catalog No. N05611G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05611G 1 g
N05615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05611G Supplier TCI America Supplier No. N05611G
Only null left

Chemical Identifiers

CAS 60835-69-0
Molecular Formula C14H19NO7
Molecular Weight (g/mol) 313.306
MDL Number MFCD00060713
InChI Key XIWRBQVYCZCEPG-UHFFFAOYSA-N
Synonym 4-nitrobenzo-15-crown-5, 4'-nitrobenzo-15-crown-5, nitrobenzo-15-crown-5, 15-nitro-2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine, 17-nitro-2,5,8,11,14-pentaoxabicyclo 13.4.0 nonadeca-1 15 ,16,18-triene, 17-nitro-2,3-benzo-1,4,7,10,13-pentaoxacyclopentadecen-2, 4-nitro-benzo-15-crown-5, xiwrbqvyczcepg-uhfffaoysa, 4'-nitrobenzo-15-crown 5-ether
PubChem CID 143747
IUPAC Name 17-nitro-2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene
SMILES C1COCCOC2=C(C=C(C=C2)[N+](=O)[O-])OCCOCCO1

Specifications

Melting Point 98°C
Color Yellow
Quantity 1 g
Formula Weight 313.31
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Nitrobenzo-15-crown 5-Ether