missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzo-15-crown-5, 99%, Thermo Scientific Chemicals

Catalog No. AAA1622506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1622506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1622506 Supplier Thermo Scientific Chemicals Supplier No. A1622506

4-Nitrobenzo-15-crown-5 is a raw material used in synthesis as well as an intemediate in chemical syntheis and pharmaceutica intemediate. It is also used to separate cations, dissolve inorganic salts into organic solvents to perform naked anion chemistry, serve as phase transfer catalysts, and chiral crown ethers have been used to resolve racemic mixtures or separate isomers by chiral chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzo-15-crown-5 is a raw material used in synthesis as well as an intemediate in chemical syntheis and pharmaceutica intemediate. It is also used to separate cations, dissolve inorganic salts into organic solvents to perform naked anion chemistry, serve as phase transfer catalysts, and chiral crown ethers have been used to resolve racemic mixtures or separate isomers by chiral chromatography.

Solubility
It is soluble in non-polar solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases

Chemical Identifiers

CAS 60835-69-0
Molecular Formula C14H19NO7
Molecular Weight (g/mol) 313.306
MDL Number MFCD00060713
InChI Key XIWRBQVYCZCEPG-UHFFFAOYSA-N
Synonym 4-nitrobenzo-15-crown-5, 4'-nitrobenzo-15-crown-5, nitrobenzo-15-crown-5, 15-nitro-2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine, 17-nitro-2,5,8,11,14-pentaoxabicyclo 13.4.0 nonadeca-1 15 ,16,18-triene, 17-nitro-2,3-benzo-1,4,7,10,13-pentaoxacyclopentadecen-2, 4-nitro-benzo-15-crown-5, xiwrbqvyczcepg-uhfffaoysa, 4'-nitrobenzo-15-crown 5-ether
PubChem CID 143747
IUPAC Name 17-nitro-2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene
SMILES C1COCCOC2=C(C=C(C=C2)[N+](=O)[O-])OCCOCCO1

Specifications

Melting Point 93°C to 95°C
Quantity 5 g
Solubility Information It is soluble in non-polar solvents.
Formula Weight 313.3
Percent Purity 99%
Chemical Name or Material 4-Nitrobenzo-15-crown-5
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only