missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzhydrazide 98.0+%, TCI America™

Catalog No. N014925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N014925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N014925G Supplier TCI America Supplier No. N014925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 636-97-5
Molecular Formula C7H7N3O3
Molecular Weight (g/mol) 181.151
MDL Number MFCD00007604
InChI Key FKZXYJYTUSGIQE-UHFFFAOYSA-N
Synonym 4-nitrobenzhydrazide, p-nitrobenzhydrazide, 4-nitrobenzoylhydrazine, p-nitrobenzohydrazide, p-nitrobenzoylhydrazide, p-nitrobenzoic hydrazide, p-nitrobenzoic acid hydrazide, p-nitrobenzoyl hydrazine, 4-nitrobenzoic acid hydrazide, 4-nitrobenzoyl hydrazide
PubChem CID 3693744
ChEBI CHEBI:66928
IUPAC Name 4-nitrobenzohydrazide
SMILES C1=CC(=CC=C1C(=O)NN)[N+](=O)[O-]

Specifications

Melting Point 210°C
Color Yellow
Quantity 25 g
Formula Weight 181.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzhydrazide
RTECSNumber DH5670000
TSCA Yes