Learn More
4-Nitrobenzhydrazide, 98+%
CAS: 636-97-5 | C7H7N3O3 | 181.151 g/mol
$124.97 - $441.33
Chemical Identifiers
| CAS | 636-97-5 |
|---|---|
| Molecular Formula | C7H7N3O3 |
| Molecular Weight (g/mol) | 181.151 |
| MDL Number | MFCD00007604 |
| InChI Key | FKZXYJYTUSGIQE-UHFFFAOYSA-N |
| Synonym | 4-nitrobenzhydrazide, p-nitrobenzhydrazide, 4-nitrobenzoylhydrazine, p-nitrobenzohydrazide, p-nitrobenzoylhydrazide, p-nitrobenzoic hydrazide, p-nitrobenzoic acid hydrazide, p-nitrobenzoyl hydrazine, 4-nitrobenzoic acid hydrazide, 4-nitrobenzoyl hydrazide |
| PubChem CID | 3693744 |
| ChEBI | CHEBI:66928 |
| IUPAC Name | 4-nitrobenzohydrazide |
| SMILES | C1=CC(=CC=C1C(=O)NN)[N+](=O)[O-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1801609
|
Thermo Scientific Chemicals
A1801609 |
10 g |
Each for $124.97
|
|
|||||
|
AAA1801618
|
Thermo Scientific Chemicals
A1801618 |
50 g |
Each for $441.33
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 636-97-5 | |
| 181.151 | |
| FKZXYJYTUSGIQE-UHFFFAOYSA-N | |
| 3693744 | |
| 4-nitrobenzohydrazide |
| C7H7N3O3 | |
| MFCD00007604 | |
| 4-nitrobenzhydrazide, p-nitrobenzhydrazide, 4-nitrobenzoylhydrazine, p-nitrobenzohydrazide, p-nitrobenzoylhydrazide, p-nitrobenzoic hydrazide, p-nitrobenzoic acid hydrazide, p-nitrobenzoyl hydrazine, 4-nitrobenzoic acid hydrazide, 4-nitrobenzoyl hydrazide | |
| CHEBI:66928 | |
| C1=CC(=CC=C1C(=O)NN)[N+](=O)[O-] |
Specifications
| 636-97-5 | |
| C7H7N3O3 | |
| 10 g | |
| 4-nitrobenzhydrazide, p-nitrobenzhydrazide, 4-nitrobenzoylhydrazine, p-nitrobenzohydrazide, p-nitrobenzoylhydrazide, p-nitrobenzoic hydrazide, p-nitrobenzoic acid hydrazide, p-nitrobenzoyl hydrazine, 4-nitrobenzoic acid hydrazide, 4-nitrobenzoyl hydrazide | |
| C1=CC(=CC=C1C(=O)NN)[N+](=O)[O-] | |
| 181.151 | |
| CHEBI:66928 | |
| ≥98% |
| ∼216°C (decomposition) | |
| MFCD00007604 | |
| 519882 | |
| FKZXYJYTUSGIQE-UHFFFAOYSA-N | |
| 4-nitrobenzohydrazide | |
| 3693744 | |
| 181.15 | |
| 4-Nitrobenzhydrazide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 211-271-1
RTECSNumber : DH5670000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only