Learn More
4-Nitrobenzenesulfonyl chloride, 97%
Description
4-Nitrobenzenesulfonyl chloride is used as an intermediate for the synthesis of pharmaceuticals like foscmprenavir, darunavir and amprenavir. It is involved in the preparation of N-nosyl-alpha-amino acid for peptide synthesis. It is also used for stereoselective α-glucosylation of partially protected carbohydrates in association with silver tri-fluoromethanesulfonate, N,N-dimethylacetamide and triethylamine. It is widely used for the synthesis of hydroxylamines of sulfadiazine, sulfamethoxazole and [Cu(bipy)2Cl](nbs) (1) (bipy = 2,2'-bipyridine, nbs = 4-nitrobenzenesulfonate).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Nitrobenzenesulfonyl chloride is used as an intermediate for the synthesis of pharmaceuticals like foscmprenavir, darunavir and amprenavir. It is involved in the preparation of N-nosyl-alpha-amino acid for peptide synthesis. It is also used for stereoselective α-glucosylation of partially protected carbohydrates in association with silver tri-fluoromethanesulfonate, N,N-dimethylacetamide and triethylamine. It is widely used for the synthesis of hydroxylamines of sulfadiazine, sulfamethoxazole and [Cu(bipy)2Cl](nbs) (1) (bipy = 2,2′-bipyridine, nbs = 4-nitrobenzenesulfonate).
Solubility
Soluble in toluene, tetrahydrofuran, methylene chloride and ethyl acetate. Insoluble in water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
| Melting Point | 76°C to 80°C |
| Boiling Point | 143°C to 144°C (1.5 mmHg) |
| Quantity | 25 g |
| UN Number | UN3261 |
| Beilstein | 746543 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in toluene,tetrahydrofuran,methylene chloride and ethyl acetate. Insoluble in water. |
| IUPAC Name | 4-nitrobenzenesulfonyl chloride |
| Formula Weight | 221.62 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | H314 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| EINECSNumber | 202-697-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only