missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzenesulfonyl chloride, 95%

Catalog No. AC415850050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC415850050 5 g Glass bottle
AC415850250 25 g Glass bottle
AC415851000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC415850050 Supplier Thermo Scientific Chemicals Supplier No. 415850050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 98-74-8 | C6H4ClNO4S | 221.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-74-8
Molecular Formula C6H4ClNO4S
Molecular Weight (g/mol) 221.61
MDL Number MFCD00007445
InChI Key JXRGUPLJCCDGKG-UHFFFAOYSA-N
Synonym 4-nitrobenzenesulfonylchloride, 4-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-nitro, 4-nitrobenzenesulphonyl chloride, 4-nitrophenylsulfonyl chloride, p-nitrophenylsulfonyl chloride, 4-nitrobenzenesulfonic acid chloride, p-nitrobenzenesulfonyl chloride, 4-nitrobenzenesulphonylchloride, 4-nitro-benzenesulfonyl chloride
PubChem CID 7404
IUPAC Name 4-nitrobenzenesulfonyl chloride
SMILES [O-][N+](=O)C1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 75°C to 80°C
Color Yellow to Brown
Boiling Point 143°C to 144°C (2.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula SO2ClC6H4NO2
Quantity 5 g
Beilstein 11, 72
Solubility Information Solubility in water: insoluble
Formula Weight 221.62
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Nitrobenzenesulfonyl chloride, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 202-697-9
TSCA TSCA

RUO – Research Use Only