Learn More
4-Nitrobenzeneboronic acid pinacol ester, 98%
CAS: 171364-83-3 | C12H16BNO4 | 249.073 g/mol
$63.19 - $211.84
Chemical Identifiers
| CAS | 171364-83-3 |
|---|---|
| Molecular Formula | C12H16BNO4 |
| Molecular Weight (g/mol) | 249.073 |
| MDL Number | MFCD02179437 |
| InChI Key | LUWACRUAJXZANC-UHFFFAOYSA-N |
| Synonym | 4,4,5,5-tetramethyl-2-4-nitrophenyl-1,3,2-dioxaborolane, 4-nitrophenylboronic acid pinacol ester, 4-nitrophenylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nitrobenzene, 4-nitrophenyl boronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-4-nitrophenyl, pubchem8088, pinacol 4-nitrophenyl boronate;, 4-nitrophenylboronate pinacol ester;, para-nitrobenzeneboronic acid pinacol ester; |
| PubChem CID | 2773555 |
| IUPAC Name | 4,4,5,5-tetramethyl-2-(4-nitrophenyl)-1,3,2-dioxaborolane |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)[N+](=O)[O-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5284803
|
Thermo Scientific Chemicals
H5284803 |
1 g |
Each for $63.19
|
|
|||||
|
AAH5284806
|
Thermo Scientific Chemicals
H5284806 |
5 g |
Each for $211.84
|
|
|||||
Description
4-Nitrobenzeneboronic acid pinacol ester is a reactant involved in synthesis of aryl azides, arylation of allylic chlorides, synthesis of RNA conjugates, alkoxycarbonylation, synthesis of mTOR and PI3K inhibitors as antitumor agents, electro oxidative synthesis of biaryls.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Nitrobenzeneboronic acid pinacol ester is a reactant involved in synthesis of aryl azides, arylation of allylic chlorides, synthesis of RNA conjugates, alkoxycarbonylation, synthesis of mTOR and PI3K inhibitors as antitumor agents, electro oxidative synthesis of biaryls.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
| 171364-83-3 | |
| 249.073 | |
| LUWACRUAJXZANC-UHFFFAOYSA-N | |
| 2773555 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)[N+](=O)[O-] |
| C12H16BNO4 | |
| MFCD02179437 | |
| 4,4,5,5-tetramethyl-2-4-nitrophenyl-1,3,2-dioxaborolane, 4-nitrophenylboronic acid pinacol ester, 4-nitrophenylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nitrobenzene, 4-nitrophenyl boronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-4-nitrophenyl, pubchem8088, pinacol 4-nitrophenyl boronate;, 4-nitrophenylboronate pinacol ester;, para-nitrobenzeneboronic acid pinacol ester; | |
| 4,4,5,5-tetramethyl-2-(4-nitrophenyl)-1,3,2-dioxaborolane |
Specifications
| 171364-83-3 | |
| C12H16BNO4 | |
| 1 g | |
| Slightly soluble in water. | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)[N+](=O)[O-] | |
| 249.073 | |
| 249.08 | |
| 4-Nitrobenzeneboronic acid pinacol ester |
| 108°C to 109°C | |
| MFCD02179437 | |
| 4,4,5,5-tetramethyl-2-4-nitrophenyl-1,3,2-dioxaborolane, 4-nitrophenylboronic acid pinacol ester, 4-nitrophenylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nitrobenzene, 4-nitrophenyl boronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-4-nitrophenyl, pubchem8088, pinacol 4-nitrophenyl boronate;, 4-nitrophenylboronate pinacol ester;, para-nitrobenzeneboronic acid pinacol ester; | |
| LUWACRUAJXZANC-UHFFFAOYSA-N | |
| 4,4,5,5-tetramethyl-2-(4-nitrophenyl)-1,3,2-dioxaborolane | |
| 2773555 | |
| 98% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only