missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzeneboronic acid pinacol ester, 98%

Catalog No. AAH5284803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5284803 1 g
AAH5284806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5284803 Supplier Thermo Scientific Chemicals Supplier No. H5284803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 171364-83-3 | C12H16BNO4 | 249.073 g/mol

4-Nitrobenzeneboronic acid pinacol ester is a reactant involved in synthesis of aryl azides, arylation of allylic chlorides, synthesis of RNA conjugates, alkoxycarbonylation, synthesis of mTOR and PI3K inhibitors as antitumor agents, electro oxidative synthesis of biaryls.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzeneboronic acid pinacol ester is a reactant involved in synthesis of aryl azides, arylation of allylic chlorides, synthesis of RNA conjugates, alkoxycarbonylation, synthesis of mTOR and PI3K inhibitors as antitumor agents, electro oxidative synthesis of biaryls.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 171364-83-3
Molecular Formula C12H16BNO4
Molecular Weight (g/mol) 249.073
MDL Number MFCD02179437
InChI Key LUWACRUAJXZANC-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-4-nitrophenyl-1,3,2-dioxaborolane, 4-nitrophenylboronic acid pinacol ester, 4-nitrophenylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nitrobenzene, 4-nitrophenyl boronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-4-nitrophenyl, pubchem8088, pinacol 4-nitrophenyl boronate;, 4-nitrophenylboronate pinacol ester;, para-nitrobenzeneboronic acid pinacol ester;
PubChem CID 2773555
IUPAC Name 4,4,5,5-tetramethyl-2-(4-nitrophenyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 108°C to 109°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 249.08
Percent Purity 98%
Chemical Name or Material 4-Nitrobenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only