missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzeneboronic acid, 95%

Catalog No. AAH2776703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2776703 1 g
AAH2776706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2776703 Supplier Thermo Scientific Chemicals Supplier No. H2776703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 24067-17-2 | C6H6BNO4 | 166.93 g/mol

It is reagent used forligand-free palladium-catalyzed Suzuki-Miyaura cross-couplings, ruthenium catalyzed direct arylation of benzylic sp3 carbons of acyclic amines, Diels-Alder or C-H activation reactions, regioselective Suzuki-Miyaura coupling and tandem palladium-catalyzed intramolecular aminocarbonylation and annulations, N-arylation of phenylurea using copper acetylacetonate catalyst, environmentally benign one-pot synthesis through a double arylation process, copper-mediated cyanation, copper-catalyzed arylations regioselective glycosylations. Suzuki couplings followed by arylations, X-ray absorption on rhodium-grafted hydrotalcite catalyst for heterogeneous 1,4-addition reaction of organoboron reagents to electron deficient olefins. It is used in Preparation of combretastatin analogs as potential antitumor agents and human immunodeficiency virus (HIV) protease inhibitors with antiviral activities against drug-resistant viruses

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is reagent used forligand-free palladium-catalyzed Suzuki-Miyaura cross-couplings, ruthenium catalyzed direct arylation of benzylic sp3 carbons of acyclic amines, Diels-Alder or C-H activation reactions, regioselective Suzuki-Miyaura coupling and tandem palladium-catalyzed intramolecular aminocarbonylation and annulations, N-arylation of phenylurea using copper acetylacetonate catalyst, environmentally benign one-pot synthesis through a double arylation process, copper-mediated cyanation, copper-catalyzed arylations regioselective glycosylations. Suzuki couplings followed by arylations, X-ray absorption on rhodium-grafted hydrotalcite catalyst for heterogeneous 1,4-addition reaction of organoboron reagents to electron deficient olefins. It is used in Preparation of combretastatin analogs as potential antitumor agents and human immunodeficiency virus (HIV) protease inhibitors with antiviral activities against drug-resistant viruses

Solubility
Slightly soluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 24067-17-2
Molecular Formula C6H6BNO4
Molecular Weight (g/mol) 166.93
MDL Number MFCD00161360
InChI Key NSFJAFZHYOAMHL-UHFFFAOYSA-N
Synonym 4-nitrophenyl boronic acid, 4-nitrobenzeneboronic acid, p-nitrophenylboronic acid, 4-nitrophenylboronicacid, p-nitrophenyl boronic acid, 4-nitro phenyl boronic acid, boronic acid, 4-nitrophenyl, boronic acid, b-4-nitrophenyl, 4-borononitrobenzene
PubChem CID 2773552
IUPAC Name (4-nitrophenyl)boronic acid
SMILES OB(O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point ∽285°C (decomposition)
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 166.93
Percent Purity 95%
Chemical Name or Material 4-Nitrobenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only