missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroaniline Hydrochloride 99.0+%, TCI America™

Catalog No. N012225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N012225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N012225G Supplier TCI America Supplier No. N012225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15873-51-5
Molecular Formula C6H7ClN2O2
Molecular Weight (g/mol) 174.58
MDL Number MFCD00054338
InChI Key LNJUVOPKIUQOQK-UHFFFAOYSA-N
PubChem CID 85156
IUPAC Name hydrogen 4-nitroaniline chloride
SMILES [H+].[Cl-].NC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 174.58
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitroaniline Hydrochloride
EINECSNumber (3)-0392
TSCA Yes