missing translation for 'onlineSavingsMsg'
Learn More

4'-Nitroacetophenone 98.0+%, TCI America™

Catalog No. p-7133625
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N011125G 25 g
N0111100G 100 g
N0111500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N011125G Supplier TCI America Supplier No. N011125G
Only null left

Chemical Identifiers

CAS 100-19-6
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD00007355
InChI Key YQYGPGKTNQNXMH-UHFFFAOYSA-N
Synonym p-nitroacetophenone, 4'-nitroacetophenone, 4-nitroacetophenone, 1-4-nitrophenyl ethanone, p-acetylnitrobenzene, paranitroacetophenone, p-nitrophenyl methyl ketone, 1-4-nitrophenyl ethan-1-one, ethanone, 1-4-nitrophenyl, pnap
PubChem CID 7487
ChEBI CHEBI:28735
IUPAC Name 1-(4-nitrophenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 79°C
Boiling Point 167°C
Quantity 25 g
Formula Weight 165.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Nitroacetophenone
EINECSNumber (9)-2076
RTECSNumber AM9627000
TSCA Yes