missing translation for 'onlineSavingsMsg'
Learn More

4'-Nitroacetophenone, 98%

CAS: 100-19-6 | C8H7NO3 | 165.15 g/mol

Supplier:  Thermo Scientific Chemicals A1387730

 View more versions of this product

Catalog No. AAA1387730

Only null left
Explore available promotions

Description

Description

4'-Nitroacetophenone is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Nitroacetophenone is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, strong alkalis.
Specifications

Specifications

4'-Nitroacetophenone
77°C to 81°C
149°C (300°F)
Characteristic
MFCD00007355
607777
Insoluble in water.
CC(=O)C1=CC=C(C=C1)[N+]([O-])=O
165.15
CHEBI:28735
98%
100-19-6
201°C to 202°C
98%
C8H7NO3
250 g
p-nitroacetophenone, 4'-nitroacetophenone, 4-nitroacetophenone, 1-4-nitrophenyl ethanone, p-acetylnitrobenzene, paranitroacetophenone, p-nitrophenyl methyl ketone, 1-4-nitrophenyl ethan-1-one, ethanone, 1-4-nitrophenyl, pnap
YQYGPGKTNQNXMH-UHFFFAOYSA-N
1-(4-nitrophenyl)ethanone
7487
165.15

Chemical Identifiers

100-19-6
165.15
YQYGPGKTNQNXMH-UHFFFAOYSA-N
7487
1-(4-nitrophenyl)ethanone
C8H7NO3
MFCD00007355
p-nitroacetophenone, 4'-nitroacetophenone, 4-nitroacetophenone, 1-4-nitrophenyl ethanone, p-acetylnitrobenzene, paranitroacetophenone, p-nitrophenyl methyl ketone, 1-4-nitrophenyl ethan-1-one, ethanone, 1-4-nitrophenyl, pnap
CHEBI:28735
CC(=O)C1=CC=C(C=C1)[N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319-H500

EINECSNumber : 202-827-4

RTECSNumber : AM9627000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only