missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-o-phenylenediamine, 98%, Thermo Scientific™

Chemical Identifiers

CAS 99-56-9
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007724
InChI Key RAUWPNXIALNKQM-UHFFFAOYSA-N
Synonym 4-nitro-o-phenylenediamine, 1,2-diamino-4-nitrobenzene, 4-nitro-1,2-phenylenediamine, 2-amino-4-nitroaniline, 3,4-diaminonitrobenzene, p-nitro-o-phenylenediamine, 1,2-benzenediamine, 4-nitro, 4-nitro-1,2-diaminobenzene, 4-nitrophenylenediamine, 4-nitro-1,2-benzenediamine
PubChem CID 5111791
ChEBI CHEBI:67116
IUPAC Name 4-nitrobenzene-1,2-diamine
SMILES NC1=CC=C(C=C1N)[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC148841000
SDS View Documents
Thermo Scientific Chemicals
148841000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC148845000
SDS View Documents
Thermo Scientific Chemicals
148845000
500g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

99-56-9
153.14
RAUWPNXIALNKQM-UHFFFAOYSA-N
5111791
4-nitrobenzene-1,2-diamine
C6H7N3O2
MFCD00007724
4-nitro-o-phenylenediamine, 1,2-diamino-4-nitrobenzene, 4-nitro-1,2-phenylenediamine, 2-amino-4-nitroaniline, 3,4-diaminonitrobenzene, p-nitro-o-phenylenediamine, 1,2-benzenediamine, 4-nitro, 4-nitro-1,2-diaminobenzene, 4-nitrophenylenediamine, 4-nitro-1,2-benzenediamine
CHEBI:67116
NC1=CC=C(C=C1N)[N+]([O-])=O

Specifications

99-56-9
100.0
97.5% min. (HPLC)
C6H7N3O2
MFCD00007724
13, 29
4-nitro-o-phenylenediamine, 1,2-diamino-4-nitrobenzene, 4-nitro-1,2-phenylenediamine, 2-amino-4-nitroaniline, 3,4-diaminonitrobenzene, p-nitro-o-phenylenediamine, 1,2-benzenediamine, 4-nitro, 4-nitro-1,2-diaminobenzene, 4-nitrophenylenediamine, 4-nitro-1,2-benzenediamine
RAUWPNXIALNKQM-UHFFFAOYSA-N
4-nitrobenzene-1,2-diamine
5111791
153.14
4-Nitro-o-phenylenediamine, 98%
97.5
Authentic
Glass bottle
O2NC6H3(NH2)2
100g
15, 6710
Solubility in water: 1.2g/L (20°C). Other solubilities: soluble in an aquous solution of hydrochloric acid, 20g/L aceton (20°C)
NC1=CC=C(C=C1N)[N+]([O-])=O
153.14
CHEBI:67116
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical ad

GHS Signal Word: Warning

EINECSNumber : 202-766-3

RTECSNumber : ST2975000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only