missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitro-DL-phenylalanine Hydrate 98.0+%, TCI America™
Supplier: TCI America N095225G
Specifications
| 4-Nitro-DL-phenylalanine Hydrate | |
| 230°C | |
| MFCD00007384 | |
| 2-Amino-3-(4-nitrophenyl)propionic Acid, H-DL-Phe(4-NO2)-OH | |
| C1=CC(=CC=C1CC(C(=O)O)N)[N+](=O)[O-].O | |
| 228.204 | |
| 210.19 | |
| Crystalline Powder |
| 207569-25-3 | |
| C9H12N2O5 | |
| 25 g | |
| OLZDHJRNUIXKLU-UHFFFAOYSA-N | |
| 2-amino-3-(4-nitrophenyl)propanoic acid;hydrate | |
| 18468090 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 207569-25-3 | |
| 228.204 | |
| OLZDHJRNUIXKLU-UHFFFAOYSA-N | |
| 18468090 | |
| C1=CC(=CC=C1CC(C(=O)O)N)[N+](=O)[O-].O |
| C9H12N2O5 | |
| MFCD00007384 | |
| 2-Amino-3-(4-nitrophenyl)propionic Acid, H-DL-Phe(4-NO2)-OH | |
| 2-amino-3-(4-nitrophenyl)propanoic acid;hydrate |
Safety and Handling
TSCA : No