Learn More
4-Nitro-alpha-toluenesulfonyl chloride, 97%
CAS: 4025-75-6 | C7H6ClNO4S | 235.638 g/mol
$103.44 - $273.62
Chemical Identifiers
| CAS | 4025-75-6 |
|---|---|
| Molecular Formula | C7H6ClNO4S |
| Molecular Weight (g/mol) | 235.638 |
| MDL Number | MFCD00034030 |
| InChI Key | CAXRPEAFHWDFTD-UHFFFAOYSA-N |
| Synonym | 4-nitrophenyl methanesulfonyl chloride, 4-nitrophenyl methanesulfonylchloride, 4-nitrobenzylsulfonyl chloride, 4-nitro-alpha-toluenesulfonyl chloride, acmc-20afqe, pubchem21404, 4-nitrobenzyl sulfonyl chloride, 4-nitrobenzyl sulphonyl chloride, 4-nitrophenylmethanesulfonylchloride, chloro 4-nitrophenyl methyl sulfone |
| PubChem CID | 2794642 |
| IUPAC Name | (4-nitrophenyl)methanesulfonyl chloride |
| SMILES | C1=CC(=CC=C1CS(=O)(=O)Cl)[N+](=O)[O-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2579403
|
Thermo Scientific Chemicals
H2579403 |
1 g |
Each for $103.44
|
|
|||||
|
AAH2579406
|
Thermo Scientific Chemicals
H2579406 |
5 g |
Each for $273.62
|
|
|||||
Description
It is used as an intermediate in organic syntheses, pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as an intermediate in organic syntheses, pharmaceuticals.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/ moisture.
Chemical Identifiers
| 4025-75-6 | |
| 235.638 | |
| CAXRPEAFHWDFTD-UHFFFAOYSA-N | |
| 2794642 | |
| C1=CC(=CC=C1CS(=O)(=O)Cl)[N+](=O)[O-] |
| C7H6ClNO4S | |
| MFCD00034030 | |
| 4-nitrophenyl methanesulfonyl chloride, 4-nitrophenyl methanesulfonylchloride, 4-nitrobenzylsulfonyl chloride, 4-nitro-alpha-toluenesulfonyl chloride, acmc-20afqe, pubchem21404, 4-nitrobenzyl sulfonyl chloride, 4-nitrobenzyl sulphonyl chloride, 4-nitrophenylmethanesulfonylchloride, chloro 4-nitrophenyl methyl sulfone | |
| (4-nitrophenyl)methanesulfonyl chloride |
Specifications
| 4025-75-6 | |
| C7H6ClNO4S | |
| 1 g | |
| Moisture sensitive | |
| Reacts with water. | |
| C1=CC(=CC=C1CS(=O)(=O)Cl)[N+](=O)[O-] | |
| 235.638 | |
| 235.64 | |
| 4-Nitro-alpha-toluenesulfonyl chloride |
| 91°C to 92°C | |
| MFCD00034030 | |
| UN3261 | |
| 4-nitrophenyl methanesulfonyl chloride, 4-nitrophenyl methanesulfonylchloride, 4-nitrobenzylsulfonyl chloride, 4-nitro-alpha-toluenesulfonyl chloride, acmc-20afqe, pubchem21404, 4-nitrobenzyl sulfonyl chloride, 4-nitrobenzyl sulphonyl chloride, 4-nitrophenylmethanesulfonylchloride, chloro 4-nitrophenyl methyl sulfone | |
| CAXRPEAFHWDFTD-UHFFFAOYSA-N | |
| (4-nitrophenyl)methanesulfonyl chloride | |
| 2794642 | |
| 97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only