missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-alpha-toluenesulfonyl chloride, 97%

Catalog No. AAH2579403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2579403 1 g
AAH2579406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2579403 Supplier Thermo Scientific Chemicals Supplier No. H2579403

CAS: 4025-75-6 | C7H6ClNO4S | 235.638 g/mol

It is used as an intermediate in organic syntheses, pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate in organic syntheses, pharmaceuticals.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/ moisture.

Chemical Identifiers

CAS 4025-75-6
Molecular Formula C7H6ClNO4S
Molecular Weight (g/mol) 235.638
MDL Number MFCD00034030
InChI Key CAXRPEAFHWDFTD-UHFFFAOYSA-N
Synonym 4-nitrophenyl methanesulfonyl chloride, 4-nitrophenyl methanesulfonylchloride, 4-nitrobenzylsulfonyl chloride, 4-nitro-alpha-toluenesulfonyl chloride, acmc-20afqe, pubchem21404, 4-nitrobenzyl sulfonyl chloride, 4-nitrobenzyl sulphonyl chloride, 4-nitrophenylmethanesulfonylchloride, chloro 4-nitrophenyl methyl sulfone
PubChem CID 2794642
IUPAC Name (4-nitrophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC=C1CS(=O)(=O)Cl)[N+](=O)[O-]

Specifications

Melting Point 91°C to 92°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 235.64
Percent Purity 97%
Chemical Name or Material 4-Nitro-alpha-toluenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only