missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylcyclohexanone, 99%

CAS: 40649-36-3 | C9H16O | 140.226 g/mol

Supplier:  Thermo Scientific Chemicals A1112614

 View more versions of this product

Catalog No. AAA1112614

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-n-Propylcyclohexanone
0.91
86°C (186°F)
1.4535
25 g
4-propylcyclohexanone, 4-n-propylcyclohexanone, cyclohexanone, 4-propyl, 4-propyl-1-cyclohexanone, 4-propylcyclohexanon, 4-propyl-cyclohexanone, pubchem14023, acmc-1arvj, 4beta-propylcyclohexanone, 4-n-propyl-cyclohexanone
CCCC1CCC(=O)CC1
140.226
140.23
40649-36-3
115°C (36 mmHg)
C9H16O
MFCD00058690
2076023
NQEDLIZOPMNZMC-UHFFFAOYSA-N
4-propylcyclohexan-1-one
142482
99%

Chemical Identifiers

40649-36-3
140.226
NQEDLIZOPMNZMC-UHFFFAOYSA-N
142482
CCCC1CCC(=O)CC1
C9H16O
MFCD00058690
4-propylcyclohexanone, 4-n-propylcyclohexanone, cyclohexanone, 4-propyl, 4-propyl-1-cyclohexanone, 4-propylcyclohexanon, 4-propyl-cyclohexanone, pubchem14023, acmc-1arvj, 4beta-propylcyclohexanone, 4-n-propyl-cyclohexanone
4-propylcyclohexan-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H227
Causes skin irritation.
Combustible liquid.

P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c

H227-H315-H319

EINECSNumber : 406-810-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only