missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propoxypiperidine, 95%

Catalog No. AAH50976MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50976MD 250 mg
AAH5097603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50976MD Supplier Thermo Scientific Chemicals Supplier No. H50976MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 88536-11-2 | C8H17NO | 143.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88536-11-2
Molecular Formula C8H17NO
Molecular Weight (g/mol) 143.23
MDL Number MFCD06248727
InChI Key VOWMRECKIQVVPP-UHFFFAOYSA-N
Synonym 4-propoxy-piperidine, 4-n-propoxypiperidine, 4-propyloxypiperidine, 4-n-propoxypiperidin, acmc-20ambz, 4-piperidyl propyl ether, 4-piperidinyl propyl ether, piperidine, 4-propoxy-, hydrochloride
PubChem CID 11492037
IUPAC Name 4-propoxypiperidine
SMILES CCCOC1CCNCC1

Specifications

Quantity 250 mg
Formula Weight 143.23
Percent Purity 95%
Chemical Name or Material 4-n-Propoxypiperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only