missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentylphenylacetylene, 97%

Catalog No. AAH6091304
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6091304 2 g
AAH6091309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6091304 Supplier Thermo Scientific Chemicals Supplier No. H6091304
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 79887-10-8 | C13H16 | 172.27 g/mol

4-n-Pentylphenylacetylene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Pentylphenylacetylene is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 79887-10-8
Molecular Formula C13H16
Molecular Weight (g/mol) 172.27
MDL Number MFCD00173884
InChI Key APGNXGIUUTWIRE-UHFFFAOYSA-N
Synonym 4-n-pentylphenylacetylene, 4-pentylphenylacetylene, 1-ethynyl-4-pentyl-benzene, 1-eth-1-ynyl-4-pentylbenzene, benzene, 1-ethynyl-4-pentyl, 4-amylphenylacetylene, 4-ethynylpentylbenzene, acmc-209pid, 4-pentylphenyl acetylene
PubChem CID 2775131
IUPAC Name 1-ethynyl-4-pentylbenzene
SMILES CCCCCC1=CC=C(C=C1)C#C

Specifications

Density 0.885
Boiling Point 172°C
Flash Point 103°C (217°F)
Refractive Index 1.523
Quantity 2 g
Formula Weight 172.27
Percent Purity 97%
Chemical Name or Material 4-n-Pentylphenylacetylene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only