missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentylbenzoyl chloride, 98%

Catalog No. p-7080635
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1024806 5 g
AAA1024814 25 g
AAA1024822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1024806 Supplier Thermo Scientific Chemicals Supplier No. A1024806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 49763-65-7 | C12H15ClO | 210.701 g/mol

4-n-Pentylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid. It is used as intermediates of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Pentylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid. It is used as intermediates of liquid crystals.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents.

Chemical Identifiers

CAS 49763-65-7
Molecular Formula C12H15ClO
Molecular Weight (g/mol) 210.701
MDL Number MFCD00000700
InChI Key FBBRKYLXMNQFQU-UHFFFAOYSA-N
Synonym benzoyl chloride, 4-pentyl, p-pentylbenzoyl chloride, 4-n-pentylbenzoyl chloride, 4-n-amylbenzoyl chloride, 4-pentyl-benzoyl chloride, 4-pentylbenzene-1-carbonyl chloride, acmc-20ambq, p-n-amyl benzoyl chloride, 4-n-pentyl benzoyl chloride, 4-n-pentyl-benzoyl chloride
PubChem CID 170812
IUPAC Name 4-pentylbenzoyl chloride
SMILES CCCCCC1=CC=C(C=C1)C(=O)Cl

Specifications

Density 1.034
Boiling Point 144°C to 145°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.53
Quantity 5 g
UN Number UN3265
Beilstein 2207887
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 210.71
Percent Purity 98%
Chemical Name or Material 4-n-Pentylbenzoyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 256-478-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only