missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentylbenzenesulfonyl chloride, 97%

Catalog No. AAA1992988
Change view
Click to view available options
Quantity:
2.5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1992988 2.5 g
AAA1992909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1992988 Supplier Thermo Scientific Chemicals Supplier No. A1992988
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 73948-18-2 | C11H15ClO2S | 246.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 73948-18-2
Molecular Formula C11H15ClO2S
Molecular Weight (g/mol) 246.75
MDL Number MFCD00173759
InChI Key XDWXZRVWJHEWMT-UHFFFAOYSA-N
Synonym 4-n-pentylbenzenesulfonyl chloride, 4-pentylbenzene-1-sulfonyl chloride, 4-t-amylbenzenesulphonyl chloride, 4-n-amylbenzenesulfonyl chloride, pubchem5685, chloro 4-pentylphenyl sulfone, 4-pentylphenylsulfonyl chloride, 4-n-amylbenzenesulphonyl choride, 4-n-pentylphenylsulfonyl chloride, 4-n-pentylbenzenesulfonly chloride
PubChem CID 144625
IUPAC Name 4-pentylbenzenesulfonyl chloride
SMILES CCCCCC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 39°C to 44°C
Quantity 2.5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 246.76
Percent Purity 97%
Chemical Name or Material 4-n-Pentylbenzenesulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only