missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentylaniline, 98%

Catalog No. AAH2656906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2656906 5 g
AAH2656914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2656906 Supplier Thermo Scientific Chemicals Supplier No. H2656906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 33228-44-3 | C11H17N | 163.264 g/mol

4-Pentylaniline is used in the synthesis of 4-[(4?-pentyl)azo]phenol (5Az-OH) by diazo-coupling reaction with phenol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Pentylaniline is used in the synthesis of 4-[(4′-pentyl)azo]phenol (5Az-OH) by diazo-coupling reaction with phenol

Solubility
Sparingly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 33228-44-3
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00007926
InChI Key DGFTWBUZRHAHTH-UHFFFAOYSA-N
Synonym benzenamine, 4-pentyl, 4-amylaniline, p-pentylaniline, p-amylaniline, unii-8roj0g9hiu, p-n-pentylaniline, 4-n-pentylaniline, 4-pentylphenylamine, 8roj0g9hiu, p-n-amylaniline
PubChem CID 93162
IUPAC Name 4-pentylaniline
SMILES CCCCCC1=CC=C(C=C1)N

Specifications

Density 0.919
Boiling Point 130°C
Flash Point >110°C (230°F)
Refractive Index 1.53
Quantity 5 g
Solubility Information Sparingly soluble in water.
Formula Weight 163.26
Percent Purity 98%
Chemical Name or Material 4-n-Pentylaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 251-409-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only