missing translation for 'onlineSavingsMsg'
Learn More

4-n-Octyloxybenzaldehyde 98.0+%, TCI America™

Catalog No. O011625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O011625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O011625G Supplier TCI America Supplier No. O011625G

Chemical Identifiers

CAS 24083-13-4
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.339
MDL Number MFCD00014136
InChI Key KVOWZHASDIKNFK-UHFFFAOYSA-N
Synonym 4-octyloxy benzaldehyde, 4-octyloxybenzaldehyde, 4-n-octyloxybenzaldehyde, p-octyloxybenzaldehyde, benzaldehyde, 4-octyloxy, p-octyloxy benzaldehyde, p-n-octoxy benzaldehyde, p-n-octyloxybenzaldehyde, acmc-209g8t
PubChem CID 90358
IUPAC Name 4-octoxybenzaldehyde
SMILES CCCCCCCCOC1=CC=C(C=C1)C=O

Specifications

Color Yellow
Boiling Point 168°C
Quantity 25 g
Formula Weight 234.34
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-n-Octyloxybenzaldehyde