missing translation for 'onlineSavingsMsg'
Learn More

4-n-Octyloxybenzaldehyde, 97%

Catalog No. p-7025291
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2069206 5 g
AAB2069214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2069206 Supplier Thermo Scientific Chemicals Supplier No. B2069206

CAS: 24083-13-4 | C15H22O2 | 234.339 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24083-13-4
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.339
MDL Number MFCD00014136
InChI Key KVOWZHASDIKNFK-UHFFFAOYSA-N
Synonym 4-octyloxy benzaldehyde, 4-octyloxybenzaldehyde, 4-n-octyloxybenzaldehyde, p-octyloxybenzaldehyde, benzaldehyde, 4-octyloxy, p-octyloxy benzaldehyde, p-n-octoxy benzaldehyde, p-n-octyloxybenzaldehyde, acmc-209g8t
PubChem CID 90358
IUPAC Name 4-octoxybenzaldehyde
SMILES CCCCCCCCOC1=CC=C(C=C1)C=O

Specifications

Density 0.982
Boiling Point 140°C to 142°C (0.1 mmHg)
Refractive Index 1.522
Quantity 5 g
Beilstein 1913284
Sensitivity Air Sensitive
Formula Weight 234.33
Percent Purity 97%
Chemical Name or Material 4-n-Octyloxybenzaldehyde
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 246-012-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only