missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-n-Octylaniline 97.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 311°C |
| Quantity | 25 g |
| Formula Weight | 205.35 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 4-n-Octylaniline |
Safety and Handling
| EINECSNumber | (3)-0206 |
| TSCA | No |